Learn More
MSA™ Passport™ VOC 2000 Organic Vapor and Oxygen Monitor Kit
Includes: VOC 2000 monitor, NiCd battery pack, 110V charger, 10 ft. (3m) coiled PTFE sampling line, 1 ft. (30cm) probe, steel belt clip, and standard packaging. VOC 2000 Monitor also available with NiCd battery pack, 110V charger, 8 in. (20.3cm) probe and pistol grip.
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
19-042-515
|
MSA™
10010864 |
Monitor with 10 ft. (3m) coiled PTFE sampling line | Monitor with 10 ft. (3m) coiled PTFE sampling line | N/A | N/A | N/A | |||
19-042-516
|
MSA™
10010865 |
Monitor with 8 in. probe | Monitor with 8 in. probe | N/A | N/A | N/A | |||
Description
Operating range of 0 to 10,000ppm VOC and 0 to 100% O2
Photo-ionization allows this compact, portable instrument to provide very low-level leak detection of volatile organic compounds (VOCs). Perfect for chemical and petroleum industry applications, it detects low concentrations of compounds such as jet fuels, methyl ethyl ketone (MEK), benzene, and toluene. An integrated electrochemical oxygen sensor also monitors oxygen levels, making the monitor appropriate for many confined-space applications as well. Download data to computer via RS-232 cable.
Compliance
MET approved for use in Class I, Division 1, Groups A, B, C, and D; Class II, Division 2, Groups E, F, and G; Class III, Division 1 and 2 hazardous locations.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.