Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Sirius™ Multigas Detector - Economy Model

Catalog No. 19123422 Shop All MSA Products
Change view
Click to view available options
Sensors:
CH4, O2, CO, H2S
LEL, O2, CO
LEL, O2, CO, H2S
Sensor Range:
0 to 100% LEL, 0 to 25% Vol O2, 0 to 200 ppm H2S
0 to 100% LEL, 0 to 25% Vol O2, 0 to 500 ppm CO
0 to 100% LEL, 0 to 25% Vol O2, 0 to 500 ppm CO, 0 to 200 ppm H2S
CH4, 0 to 25% Vol O2, 0 to 500 ppm CO, 0 to 200 ppm H2S
Sensor Resolution:
1% LEL, 0.1% Vol O2, 1 ppm CO, 1 ppm H2S
CH4, 0.1% Vol O2, 1 ppm CO, 1 ppm H2S
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sensors Sensor Range Sensor Resolution
19-123-422 - - -
19-123-408 LEL, O2, CO, H2S 0 to 100% LEL, 0 to 25% Vol O2, 0 to 500 ppm CO, 0 to 200 ppm H2S 1% LEL, 0.1% Vol O2, 1 ppm CO, 1 ppm H2S
19-123-409 - 0 to 100% LEL, 0 to 25% Vol O2, 0 to 200 ppm H2S -
19-123-410 LEL, O2, CO 0 to 100% LEL, 0 to 25% Vol O2, 0 to 500 ppm CO -
19-123-411 LEL, O2, CO, H2S 0 to 100% LEL, 0 to 25% Vol O2, 0 to 500 ppm CO, 0 to 200 ppm H2S 1% LEL, 0.1% Vol O2, 1 ppm CO, 1 ppm H2S
19-123-421 - - -
19-123-420 CH4, O2, CO, H2S CH4, 0 to 25% Vol O2, 0 to 500 ppm CO, 0 to 200 ppm H2S CH4, 0.1% Vol O2, 1 ppm CO, 1 ppm H2S
19-123-423 - - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-123-422 Supplier MSA™ Supplier No. 10051187

Includes

Economy Models include: Monitor, sensors and batteries

Combines PID functionality with a four-gas monitor

Integrates high-performance photoionization detector sensor with a multigas detector for up to four gases. Users are able to simultaneously monitor for volatile organic compounds (VOCs) with low vapor pressures while measuring for combustible, toxic, and oxygen-deficient atmospheres within one reliable, easy-to-use, durable unit.

Features:
•     Measures VOC readings first for greater accuracy of ±10% or 2ppm over full range
•     Best in class humidity performance
•     Notifies failure or removal of lamp or ion chamber
•     Features an over range indicator, external exhaust, and multidirectional audio and visual alarms
•     Lamps are easy to replace and clean

For use in:
•     Leak detection
•     Perimeter monitor
•     Spill delineation
•     Remediation
•     Arson investigation
•     Diesel fuel TVL monitoring (Marine Chemist)
•     Decontamination
•     PPE Determination

Compliance

Meets 47CFR part 15. UL913 6th edition and CSA 22.2 No. 157 for use in hazardous locations Class I, Divisions I, Groups A, B, C, and D. Temperature class T3C. Meets Canadian combustible performance standard CSA 22.2 No. 152 for methane only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.