Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mtTFA Antibody (18G102B2E11), Novus Biologicals™
SDP

Catalog No. NBP171648SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP171648SS 0.025 mL
NBP171648 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP171648SS Supplier Novus Biologicals Supplier No. NBP171648SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 9 publications

mtTFA Monoclonal specifically detects mtTFA in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated, Knockdown Validated.

Specifications

Antigen mtTFA
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay, KnockDown
Classification Monoclonal
Clone 18G102B2E11
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western 1:20, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1-5 ug/ml, Immunohistochemistry-Paraffin 1:200, Knockout Validated reported in scientific literature (PMID 32001952), KnockDown Validated
Gene Accession No. Q00059
Gene Alias mitochondrial transcription factor A, MtTF1, mtTFA, TCF-6, TCF6L1, TCF6L2Mitochondrial transcription factor 1, TCF6TCF6L3, Transcription factor 6, Transcription factor 6-like 2, Transcription factor 6-like 2 (mitochondrial transcription factor), transcription factor A, mitochondrial
Gene Symbols TFAM
Host Species Mouse
Immunogen Human mtTFA [Swiss-Prot# Q00059]
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7019
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.