Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MUC5B Monoclonal Antibody (C10)
Mouse Monoclonal Antibody
$213.50 - $522.50
Specifications
Antigen | MUC5B |
---|---|
Clone | C10 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541641
|
Invitrogen™
MA541641 |
200 μL |
Each for $522.50
|
|
|||||
PIMA543650
|
Invitrogen™
MA543650 |
20 μL |
Each for $213.50
|
|
|||||
Description
MUC5B Monoclonal Antibody for Western Blot
Mucins (MUCs) are heavily glycosylated glycoproteins that are synthesized either as membrane-bound or secreted proteins. Mucins are noteworthy because they have an aberrant expression profile in various malignancies. Mucins normally play a protective role for epithelial tissues and are expressed in the gastrointestinal, respiratory, and genito-urinary tracts. Mucins may also be involved in the renewal and differentiation of the epithelium, and the modulation of cell adhesion and cell signaling. MUC5B is thought to play a vital role in maintaining oral health, and may also be involved in respiratory and oral disease.2 MUC5B expression has been studied in gastric carcinoma as well as chronic obstructive pulmonary disease.Specifications
MUC5B | |
1 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9HC84 | |
727897 | |
Recombinant protein Mucin 5 Subtype B. The antigen corresponds to amino acid range 5653-5762 of the target protein. | |
Antibody |
C10 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2b κ | |
Human | |
MUC5B | |
Cervical mucin; cervical mucin MUC5B; high molecular weight salivary mucin MG1; MG1; MUC5; MUC5B; MUC-5B; MUC9; mucin 5, subtype B, tracheobronchial; mucin 5B, oligomeric mucus/gel-forming; Mucin-5 subtype B, tracheobronchial; Mucin-5B; sublingual gland mucin | |
MUC5B | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title