Learn More
Mucin 2 (MUC2) Ab-2 Mouse Monoclonal Antibody, Epredia™
Deliver accurate, reproducible results with the Epredia™ Mucin 2 (MUC2) Ab-2, Mouse Monoclonal Antibody.
Specifications
Antigen | Mucin 2 (MUC2) Ab-2 |
---|---|
Clone | M53 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Form | Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Concentration | Quantity | Price | Quantity & Availability | |||
MS1037P1
|
Epredia
MS1037P1 |
Concentrated | 200μg/mL | 500 μL | N/A | N/A | N/A | ||
MS1037P
|
Epredia
MS1037P |
Concentrated | 200μg/mL | 1 mL | N/A | N/A | N/A | ||
MS1037P1ABX
|
Epredia
MS1037P1ABX |
Concentrated | 1mg/mL | 100 μL | N/A | N/A | N/A | ||
MS1037PABX
|
Epredia
MS1037PABX |
Concentrated | 1mg/mL | 200 μL | N/A | N/A | N/A | ||
MS1037R1
|
Epredia
MS1037R1 |
Prediluted | — | 1 mL | N/A | N/A | N/A | ||
MS1037R7
|
Epredia
MS1037R7 |
Prediluted | — | 7 mL | N/A | N/A | N/A | ||
MS1037RQ
|
Epredia
MS1037RQ |
Ready-to-Use | — | 12 mL | N/A | N/A | N/A | ||
Description
Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.Host Species: Mouse
Clone: M53
Isotype: IgG2a/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant protein encoding five tandem repeats of the human MUC2 apomucin
Molecular Weight: 520kDa
Positive Control: Small intestine
Cellular Localization: Cytoplasmic and cell surface
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Mucin 2 (MUC2) Ab-2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Recombinant protein encoding 5 tandem repeats of the human MUC2 apomucin |
M53 | |
Monoclonal | |
Mouse | |
Cancer and Tumor Biology | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.