Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MuRF1/TRIM63 Antibody, Novus Biologicals™
SDP

Catalog No. NB018832 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018832 0.025 mg
NBP21106105 0.05 mg
NBP211061 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB018832 Supplier Novus Biologicals Supplier No. NBP2110610.025MG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MuRF1/TRIM63 Polyclonal specifically detects MuRF1/TRIM63 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen MuRF1/TRIM63
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.50 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:500
Formulation Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide
Gene Alias EC 6.3.2.-, IRFMURF2, Iris RING finger protein, MuRF1, MuRF-1, muscle specific ring finger protein 2, Muscle-specific RING finger protein 1, RING finger protein 28FLJ32380, RNF28E3 ubiquitin-protein ligase TRIM63, SMRZMURF-1, Striated muscle RING zinc finger protein, tripartite motif containing 63, tripartite motif-containing 63, Tripartite motif-containing protein 63
Gene Symbols TRIM63
Host Species Rabbit
Immunogen Synthetic peptide taken from human MURF1 protein taken from within the C-terminal domain of MURF1 corresponding to amino acid sequence. The peptide was conserved in several other species. The selected peptide was post-synthetically modified to achieve highest antigenicity before coupling to carrier protein using hetero bifunctional cross linker for immunogen preparation
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 84676
Test Specificity Q969Q1 The antibody labels a strong diffuse band of MURF1of 41 kDa in PC-MURF1samples and in several muscle cell lines. The antibody also labels a diffuse band of 73kDa. The identity of this band is not known.
Target Species Mouse, Rat, Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.