Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myogenin Antibody (5FD) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 9 publications
$206.00
Specifications
Antigen | Myogenin |
---|---|
Clone | 5FD |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 2 ug/mL, Immunohistochemistry 1:20 - 1:1000, Immunocytochemistry/Immunofluorescence 5 ug/mL, Immunohistochemistry-Paraffin 20 - 40 ug/mL, Immunohistochemistry-Frozen 10 - 20 ug/mL |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056510T
![]() |
Novus Biologicals
NB10056510SS |
0.025 mg |
Each for $206.00
|
|
|||||
NB10056510
![]() |
Novus Biologicals
NB10056510 |
0.1 mg | N/A | N/A | N/A | ||||
Description
Myogenin Monoclonal specifically detects Myogenin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Myogenin | |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Stem Cell Markers, Transcription Factors and Regulators | |
P15173 | |
4656 | |
A recombinant protein corresponding to amino acids 30-224 of rat myogenin was used as immunogen (NP_002470). | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
5FD | |
Western Blot 2 ug/mL, Immunohistochemistry 1:20 - 1:1000, Immunocytochemistry/Immunofluorescence 5 ug/mL, Immunohistochemistry-Paraffin 20 - 40 ug/mL, Immunohistochemistry-Frozen 10 - 20 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
BHLHC3, bHLHc3Myogenic factor-4; myogenin, Class C basic helix-loop-helix protein 3, myf-4, MYF4myogenin, Myogenic factor 4, MYOGENIN, myogenin (myogenic factor 4) | |
MYOG | |
IgG1 | |
Protein G purified | |
The 5FD clone has been highly published; users are encouraged to consult the scientific literature for additional application information. In Rh-30, a 34 kDa band should be observed. The can be used for gel shift assays. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title