Learn More
Epredia™ Lab Vision™ Myosin, Skeletal Muscle Ab-2, Mouse Monoclonal Antibody
Provides accurate, reproducible results with the Epredia™ Myosin, Skeletal Muscle Ab-2, Mouse Monoclonal Antibody.
$2330.00
Specifications
Antigen | Myosin, Skeletal Muscle Ab-2 |
---|---|
Clone | MYSN02; same as MY-32 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
MS1236S1
|
Epredia™
MS1236S1 |
Concentrated | 500 μL |
Each for $2,330.00
|
|
||||
MS1236S0
|
Epredia™
MS1236S0 |
Concentrated | 100 μL | N/A | N/A | N/A | |||
MS1236S
|
Epredia™
MS1236S |
Concentrated | 1 mL | N/A | N/A | N/A | |||
MS1236R7
|
Epredia™
MS1236R7 |
Prediluted | 7 mL | N/A | N/A | N/A | |||
Description
Myosin is a 480kDa protein known to interact with actin in muscle and in non-muscle cells. It contains two identical heavy chains (200kDa each) and four light chains (15-26kDa). Myosin molecules consist of two major regions: tails (rod) and heads; they aggregate into filaments through the tail region and interact with actin and with ATP through the head region. Multiple forms of myosin heavy chains exist for each muscle type-skeletal, cardiac, smooth and non-muscle isomyosin forms exist in different types of skeletal muscle, depending on the physiological function of the muscle. These are designated at type I (slow-twitch) and type II (fast-twitch).Host Species: Mouse
Clone: MYSN02; same as MY-32
Isotype: IgG1
Species Reactivity: Human, Cow, Mouse, Rat, Rabbit, Chicken and Guinea pig.
Epitope: Not determined
Immunogen: Rabbit muscle myosin
Molecular Weight: 480kDa
Positive Control: Skeletal muscle
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Myosin, Skeletal Muscle Ab-2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Bovine, Chicken, Guinea Pig, Human, Mouse, Rabbit, Rat | |
IgG1 |
MYSN02; same as MY-32 | |
Monoclonal | |
Mouse | |
Muscle Biology | |
Rabbit muscle myosin | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.