Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dioctyl sulfosuccinate, sodium salt, 96%
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Plastic Bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Acidity | 2.5 max. (on solids basis) |
| Melting Point | 153.0°C to 157.0°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 95% min |
| Packaging | Plastic Bottle |
| Quantity | 1 kg |
| Beilstein | 04, IV, 114 |
| Fieser | 15,149 |
| Merck Index | 15, 3446 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye damage. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/e |
| EINECSNumber | 209-406-4 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction