Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NALP4 Antibody, Novus Biologicals™
SDP

Catalog No. p-5082529 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056156 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056156 Supplier Novus Biologicals Supplier No. NB10056156
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

NALP4 Polyclonal specifically detects NALP4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen NALP4
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000
Formulation Whole antisera with 0.02% Sodium Azide
Gene Alias Cancer/testis antigen 58, CLR19.5, CT58PYRIN and NACHT-containing protein 2, FLJ32126, NACHT, leucine rich repeat and PYD containing 4, NACHT, LRR and PYD domains-containing protein 4, NALP4NACHT, LRR and PYD containing protein 4, NLR family, pyrin domain containing 4, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 4, PAN2Ribonuclease inhibitor 2, PYPAF4PAAD and NACHT-containing protein 2, RNH2PYRIN-containing APAF1-like protein 4
Gene Symbols NLRP4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 139-157 (FAPKETGKQPRTVIIQGPQ) of human NALP4/PAN2/PYPAF4 was used as immunogen; GenBank no. gi 17064172 gb AAL35293.1 AF442488_1
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147945
Test Specificity NALP4
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.