Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Native Lipoteichoic Acid Mouse anti-Bacteria, Clone: mAb 55, Novus Biologicals™
SDP

Catalog No. p-7200009 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018700 0.025 mg
NBP16014601 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018700 Supplier Novus Biologicals Supplier No. NBP1601460.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Native Lipoteichoic Acid Monoclonal specifically detects Native Lipoteichoic Acid in Bacteria samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunoassay.

Specifications

Antigen Native Lipoteichoic Acid
Applications Immunofluorescence, Immunohistochemistry, Western Blot, Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone mAb 55
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 1:250, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay 1.2 ug/ml in PBS
Formulation PBS. 1 ml >200 ug/ml) culture medium with a low endotoxin level with 0.02% Sodium Azide
Gene Alias LTA
Host Species Mouse
Immunogen Microbial mixture of Streptococcus sobrims HG961, HG962, HG970, and HG977
Purification Method Sterile filtered
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The specificity is labled not applicable.
Target Species Other
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.