Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDFIP1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB341502 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341502 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341502 Supplier Novus Biologicals Supplier No. NBP282012MFV500
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NDFIP1 Polyclonal antibody specifically detects NDFIP1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen NDFIP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Breast cancer-associated protein SGA-1M, MGC10924, N4WBP5Putative NF-kappa-B-activating protein 164, Nedd4 family interacting protein 1, NEDD4 family-interacting protein 1, NEDD4 WW domain-binding protein 5, Putative MAPK-activating protein PM13, Putative NFKB and MAPK-activating protein
Host Species Rabbit
Immunogen NDFIP1 antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human NDFIP1. The immunogen is located within the first 50 amino acids of NDFIP1. Amino Acid Squence: EEESGEPEQAAGDAPPPYS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 80762
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.