Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEK11 Antibody (OTI4H5), PE, Novus Biologicals™
SDP

Catalog No. NB287676 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287676 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287676 Supplier Novus Biologicals Supplier No. NBP272933PE
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NEK11 Monoclonal antibody specifically detects NEK11 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen NEK11
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI4H5
Conjugate PE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias EC 2.7.11, EC 2.7.11.1, FLJ23495, Never in mitosis A-related kinase 11, NIMA (never in mitosis gene a)- related kinase 11, NimA-related protein kinase 11, serine/threonine-protein kinase Nek11
Host Species Mouse
Immunogen Full length human recombinant protein of human NEK11 (NP_079076) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 79858
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.