Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nanog Antibody (5A10) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7106173 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104320SS 0.025 mL
NBP104320 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP104320SS Supplier Novus Biologicals Supplier No. NBP104320SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 6 publications

Nanog Monoclonal specifically detects Nanog in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Nanog
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5A10
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100
Formulation PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9H9S0
Gene Alias FLJ12581, FLJ40451, hNanog, homeobox protein NANOG, Homeobox transcription factor Nanog, homeobox transcription factor Nanog-delta 48, Nanog homeobox
Gene Symbols NANOG
Host Species Mouse
Immunogen Recombinant human Nanog (1-154aa) purified from E. coli (NP_079141).
Molecular Weight of Antigen 35 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Embryonic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 79923
Test Specificity Anti-human Nanog mAb, clone 5A10, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Nanog protein.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.