Learn More
Eppendorf™ Scientific Test Tube Racks
Adjustable angle and tube height
$690.90 - $856.68
Catalog Number | Mfr. No. | Size | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Diameter (Metric) | Price | Quantity & Availability | ||||
14-285-703
|
Eppendorf™
M12890002 |
Small | 12 to 15 mm |
Each for $811.84
|
|
||||
14-285-702
|
Eppendorf™
M12890001 |
Small | 8 to 11 mm |
Each for $810.66
|
|
||||
14-285-704
![]() |
Eppendorf™
M12890003 |
Small | 15 to 18 mm |
Each for $810.66
|
![]() |
||||
14-285-705
|
Eppendorf™
M12890004 |
Small | 18 to 21 mm |
Each for $721.35
|
|
||||
14-285-706
|
Eppendorf™
M12890005 |
Small | 22 to 26 mm |
Each for $690.90
|
|
||||
14-285-708
|
Eppendorf™
M12890010 |
Medium | 8 to 11 mm |
Each for $856.68
|
|
||||
14-285-709
|
Eppendorf™
M12890020 |
Medium | 12 to 15 mm |
Each for $856.68
|
|
||||
14-285-710
|
Eppendorf™
M12890030 |
Medium | 15 to 18 mm |
Each for $856.68
|
|
||||
14-285-707
![]() |
Eppendorf™
M12890006 |
Small | 26 to 30 mm |
Each for $810.66
|
![]() |
||||
14-285-711
|
Eppendorf™
M12890040 |
Medium | 18 to 21 mm |
Each for $762.30
|
|
||||
14-285-713
|
Eppendorf™
M12890060 |
Medium | 26 to 30 mm |
Each for $815.38
|
|
||||
14-285-712
|
Eppendorf™
M12890050/DEL |
Medium | 22 to 26 mm | N/A | N/A | N/A | |||
Description
- Inserts to help retain tubes and prevent them from spinning in the rack
- Finger-tight knobs to quickly adjust angle, or release the rack from the base plate, for easy transport to the bench
- Stainless-steel
Specifications
Test Tube Rack | |
Stainless Steel |