Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nextteq™ Gastec™ Detector Tubes: Formaldehyde
Shop All Nextteq Products

Click to view available options
Components Measured:
Formaldehyde
Formaldehyde Low Range
Measurement Range:
0.05 to 1
0.1 to 40
2 to 100
Response Time:
1.5 to 7.5 min.
30 sec. to 1.5 min.
7.5 min.
No. of Strokes:
0.5 to 2
1 to 5
5
Description
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.
Specifications
Specifications
| Components Measured | Formaldehyde |
| Measurement Range | 0.1 to 40 |
| No. of Strokes | 1 to 5 |
| Quantity | 10 |
| Response Time | 1.5 to 7.5 min. |
| Temperature Range (English) | 32°F to 104°F |
| Temperature Range (Metric) | 0°C to 40°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction