Learn More
Description
Specifications
Specifications
| Antigen | NG2/MCSP |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | LHM 2 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Flow Cytometry, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:100, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen 1:200, Flow (Cell Surface) 1 - 2.5 ug/mL |
| Gene Accession No. | Q6UVK1 |
| Gene Alias | chondroitin sulfate proteoglycan 4, chondroitin sulfate proteoglycan 4 (melanoma-associated), EC 3.6.3, HMW-MAA, MCSPChondroitin sulfate proteoglycan NG2, MCSPGMelanoma chondroitin sulfate proteoglycan, MEL-CSPG, MSK16Melanoma-associated chondroitin sulfate proteoglycan, NG2EC 2.7.8 |
| Gene Symbols | CSPG4 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
