Learn More
Description
Specifications
Specifications
| Antigen | nkx6.2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | GTXhomeobox 6B, Homeobox protein NK-6 homolog B, homeobox protein Nkx-6.2, MGC126684, NK homeobox family 6, B, NK6 homeobox 2, NK6 transcription factor related, locus 2, NK6 transcription factor related, locus 2 (Drosophila), NKX6.1, NKX6.2, NKX6Bglial and testis-specific homeobox protein |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human nkx6.2 (NP_796374). Peptide sequence PLAALHNMAEMKTSLFPYALQGPAGFKAPALGGLGAQLPLGTPHGISDIL |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
