Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRC3 Antibody (Eowyn-1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB013215 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013215 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013215 Supplier Novus Biologicals Supplier No. NBP280084
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NLRC3 Monoclonal specifically detects NLRC3 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen NLRC3
Applications Western Blot
Classification Monoclonal
Clone Eowyn-1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS containing 10% glycerol
Gene Alias CARD15-Like Protein, Caterpiller Protein 16.2, CLR16.2, Leucine Rich Repeat And CARD Domain Containing 3, NLR Family, CARD Domain Containing 3, NOD3, NOD-Like Receptor C3, Nucleotide-Binding Oligomerization Domain Protein 3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And CARD Domain Containing 3, Protein NLRC3
Gene Symbols NLRC3
Host Species Mouse
Immunogen Recombinant mouse NLRC3 (card domain).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 197358
Test Specificity Detects overexpressed mouse NLRC3. Doest not detect human NLRC3. Although NLRC3 (mouse), mAb (Eowyn-1) is strong and specific (see figure), the endogenous protein has not been detected yet with this antibody (using NLRC3 KO versus WT immune cells). NLRC3 protein is probably expressed at very low levels or its translation/ stability is triggered by stimuli still to be discovered .
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.