Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRP3/NALP3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7109110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177080SS 0.025 mg
NBP177080 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP177080SS Supplier Novus Biologicals Supplier No. NBP177080SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 23 publications

NLRP3/NALP3 Polyclonal specifically detects NLRP3/NALP3 in Human, Mouse, E. coli samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, SDS-Page.

Specifications

Antigen NLRP3/NALP3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunoprecipitation reported in scientific literature (PMID 31551961), Immunohistochemistry-Paraffin 1:200, SDS-Page reported in scientific literature (PMID 31551961)
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q96P20
Gene Alias AII, AII/AVP, Angiotensin/vasopressin receptor AII/AVP-like, AVP, C1orf7, Caterpiller protein 1.1, CLR1.1, Cold autoinflammatory syndrome 1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, Familial cold autoinflammatory syndrome, FCU, Leucine-rich repeat-, and PYD-containing protein 3, Muckle-Wells syndrome, MWS, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, NALP3, NLR family, pyrin domain containing 3, NLRP3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 3, PYPAF1, PYRIN-containing APAF1-like protein 1
Gene Symbols NLRP3
Host Species Rabbit
Immunogen NLRP3/NALP3 Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human NALP3. The immunogen is located within amino acids 120 - 170 of NALP3.
Molecular Weight of Antigen 110 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Inflammation, Neurodegeneration, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 114548
Target Species Human, Mouse, E. coli
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.