Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | PSMC1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP191476
![]() |
Novus Biologicals
NBP191476 |
100 μL |
Each for $487.50
|
|
|||||
NBP19147620
![]() |
Novus Biologicals
NBP19147620UL |
20 μL | N/A | N/A | N/A | ||||
Description
PSMC1 Polyclonal specifically detects PSMC1 in Mouse samples. It is validated for Western Blot, Simple Western.Specifications
PSMC1 | |
Polyclonal | |
Rabbit | |
P62192 | |
5700 | |
Synthetic peptide corresponding to a region of Mouse Psmc1 (NP_032973). Peptide sequence ELFRVAEEHAPSIVFIDEIDAIGTKRYDSNSGGEREIQRTMLELLNQLDG. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
26S protease regulatory subunit 4, 26S proteasome AAA-ATPase subunit RPT2, MGC24583, P26S4, p56, proteasome (prosome, macropain) 26S subunit, ATPase, 1, proteasome 26S ATPase subunit 1, Proteasome 26S subunit ATPase 1, proteasome 26S subunit, ATPase, 1, S4MGC8541 | |
PSMC1 | |
IgG | |
48 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title