Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLHL12/C3IP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | KLHL12/C3IP1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18240420
![]() |
Novus Biologicals
NBP18240420UL |
20 μL |
Each for $206.00
|
|
|||||
NBP182404
![]() |
Novus Biologicals
NBP182404 |
100 μL |
Each for $487.50
|
|
|||||
Description
KLHL12/C3IP1 Polyclonal specifically detects KLHL12/C3IP1 in Rat samples. It is validated for Western Blot.Specifications
KLHL12/C3IP1 | |
Polyclonal | |
Rabbit | |
NP_714952 | |
59349 | |
Synthetic peptide towards C3IP1. Peptide sequence DIMTNTHAKSILNSMNSLRKSNTLCDVTLRVEQKDFPAHRIVLAACSDYF. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
C3IP1kelch-like protein 12, CUL3-interacting protein 1, DKIR, FLJ27152, kelch-like 12 (Drosophila), kelch-like protein C3IP1 | |
KLHL12 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title