Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAA11 Antibody, Novus Biologicals™
SDP

Catalog No. p-7110151 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP191327 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP191327 Supplier Novus Biologicals Supplier No. NBP191327
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NAA11 Polyclonal specifically detects NAA11 in Rat samples. It is validated for Western Blot.

Specifications

Antigen NAA11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_001019913
Gene Alias ARD1 homolog B, ARD1B, ARD2, hARD2, human arrest defective 2, MGC10646, N(alpha)-acetyltransferase 11, NatA catalytic subunit, NatA catalytic subunit, N-terminal acetyltransferase complex ARD1 subunit homolog B
Gene Symbols NAA11
Host Species Rabbit
Immunogen The specific Immunogen is proprietary information. Peptide sequence SAKYVSLHVRKSNRAALHLYSNTLNFQVSEVEPKYYADGEDAYAMKRDLA.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84779
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.