Learn More
Description
Specifications
Specifications
| Antigen | Cytomegalovirus |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | DDG9 and CCH2 |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:10-1:25, Immunohistochemistry-Paraffin 1:10-1:25 |
| Formulation | Tissue culture supernatant with 0.05% Sodium Azide |
| Gene Alias | CMV; HHV 5; HHV5; Human herpesvirus 5 |
| Host Species | Mouse |
| Immunogen | BALB/C mice were immunized with cytomegalovirus infected cell lysate. This antibody is a cocktail of DDG9 and CCH2. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
