Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOC4L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | NOC4L |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18046420
![]() |
Novus Biologicals
NBP18046420UL |
20 μL |
Each for $206.00
|
|
|||||
NBP180464
![]() |
Novus Biologicals
NBP180464 |
100 μL |
Each for $487.50
|
|
|||||
Description
NOC4L Polyclonal specifically detects NOC4L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NOC4L | |
Polyclonal | |
Rabbit | |
NP_076983 | |
79050 | |
Synthetic peptide directed towards the C terminal of human NOC4L. Peptide sequence CRVLVHRPHGPELDADPYDPGEEDPAQSRALESSLWELQALQRHYHPEVS. | |
Primary |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
MGC3162, NET49, NOC4, NOC4 protein homolog, NOC4-like protein, nucleolar complex associated 4 homolog (S. cerevisiae), nucleolar complex protein 4 homolog, Nucleolar complex-associated protein 4-like protein | |
NOC4L | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title