Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Nonradioactive Phosphatase Assay Systems
Shop All Promega Corporation Products
Click to view available options
Product Type:
Ser or Thr system
Description
- Absorbance-Based Detection of Phosphatase Activity
- Choose from serine/threonine or tyrosine phosphatase assays
- Use with purified samples, crude cell or tissue extracts
- Convenient alternative to radioactive phosphatase assays
Specifications
Specifications
For Use With (Application) | Can be used with partially fractionated and purified samples as well as in crude cell or tissue extracts |
Content And Storage | 2 to 10°C |
Product Type | Ser or Thr system |
Quantity | 96 Reactions |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction