Learn More
MilliporeSigma™ Novagen™ KOD Hot Start Master Mix
GSA_VA
For convenient high fidelity PCR
$347.50 - $1348.00
Specifications
Content And Storage -20°C
Product Type KOD Hot Start Master Mix
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
71-842-3
GSA_VA
View Documents
MilliporeSigma™
718423
100 rxn
Each for $347.50
Only null left
Add to Cart
71-842-4
View Documents
MilliporeSigma™
718424
500 rxn
Each for $1,348.00
Only null left
Add to Cart
Description
Description
  • High accuracy, yield, and processivity of commercially available proofreading DNA polymerases
  • Amplifies genomic DNA templates up to 12 kbp
  • Amplifies plasmid and lambda DNA template up to 21 kbp
  • Eliminates mispriming and primer-dimer formation
  • Convenient ambient-temperature setup compatible with automation
  • KOD Hot Start Buffer ensures optimal PCR performance over wide range of targets
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
-20°C
KOD Hot Start Master Mix
Videos
SDS
Documents
Documents
Product Certifications
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.