Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Chicken Universal Protein Whole Tissue Lysate (Adult Whole Normal)
Shop All R&D Systems Products

Click to view available options
Quantity:
2 Units
Description
The Chicken Universal Protein Whole Tissue Lysate (Adult Whole Normal) is derived from chicken.
Specifications
Specifications
| Tissue | Universal |
| Quantity | 2 Units |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Test Specificity | Universal Protein Lysate: Chicken Normal Tissues |
| Cellular Fraction | Whole |
| Tissue Condition | Normal |
| Cross Reactivity | Chicken |
| Host Species | Chicken |
| Product Type | Chicken Universal Protein Whole Tissue Lysate (Adult Normal) |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Show More |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction