Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Lung Whole Tissue Lysate (Adult Whole Liver Cirrhosis)
SDP

Catalog No. NB82059329 Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
The Human Lung Whole Tissue Lysate (Adult Whole Liver Cirrhosis) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.

Specifications

For Use With (Application) Western Blot
Tissue Respiratory
Quantity 1 mg
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Total Protein - Liver Cirrhosis: Lung
Cellular Fraction Whole
Tissue Condition Liver Cirrhosis
Cross Reactivity Human
Host Species Human
Concentration 5 mg/mL
Product Type Human Lung Whole Tissue Lysate (Adult Liver Cirrhosis)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.