Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Brain Whole Tissue Lysate (Adult Whole Normal)
SDP

Catalog No. NB82059657 Shop All R&D Systems Products
Click to view available options
:
1mg

Catalog No. NB82059657

Supplier: Novus Biologicals™ NB82059657

Only null left
Add to Cart
Add to Cart
The Mouse Brain Whole Tissue Lysate (Adult Whole Normal) is derived from mouse. It has been validated for the following applications: Western Blot, Immunoprecipitation, SDS-Page.

Specifications

Tissue Brain
For Use With (Application) Immunoprecipitation, SDS-Page, Western Blot
Quantity 1 mg
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Purity Protein
Test Specificity Total Protein - Mouse Normal Tissue: Brain
Cellular Fraction Whole
Tissue Condition Normal
Cross Reactivity Mouse
Host Species Mouse
Concentration 5 mg/mL
Product Type Mouse Brain Whole Tissue Lysate (Adult Normal)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.