Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ MYH Overexpression Lysate
Shop All Novus Biologicals Products

Click to view available options
Quantity:
0.1 mg
Description
The MYH Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_141638. The protein contains a C-terminal DDK tag. |
For Use With (Application) | Western Blot |
Quantity | 0.1 mg |
Storage Buffer | RIPA buffer |
Purity | Protein |
Test Specificity | Homo sapiens MYH, mRNA. |
Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
Research Category | Base Excision Repair, Cancer, DNA Repair |
Target | MYH |
Synonym | CYP2C, EC 3.2.2, EC 3.2.2.-, hMYH, MGC4416, mutY (E. coli) homolog, MutY homolog, mutY homolog (E. coli), Muyth, MYHA/G-specific adenine DNA glycosylase |
Show More |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction