Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Description
The SLC22A8 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
    
                
    
            Specifications
Specifications
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC2556 and based on accession number NM_4254. The protein contains a C-terminal DDK tag. | 
| For Use With (Application) | Western Blot | 
| Storage Buffer | RIPA buffer | 
| Purity | Protein | 
| Test Specificity | Homo sapiens solute carrier family 22 (organic anion transporter), member 8 (SLC22A8), mRNA. | 
| Content And Storage | -80°C. Avoid freeze-thaw cycles. | 
| Protein State | Native | 
For Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                