Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nSMase Antibody, Novus Biologicals™
SDP

Catalog No. p-7108392 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP159937 100 μL
NBP15993720 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP159937 Supplier Novus Biologicals Supplier No. NBP159937
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

nSMase Polyclonal specifically detects nSMase in Human samples. It is validated for Western Blot.

Specifications

Antigen nSMase
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. O60906
Gene Alias EC 3.1.4.12, ISC1, lyso-PAF-PLC, Lyso-platelet-activating factor-phospholipase C, Neutral sphingomyelinase, N-SMase, NSMASE, NSMASE1, sphingomyelin phosphodiesterase 2, sphingomyelin phosphodiesterase 2, neutral membrane (neutral sphingomyelinase)
Gene Symbols SMPD2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SMPD2(sphingomyelin phosphodiesterase 2, neutral membrane (neutral sphingomyelinase)) The peptide sequence was selected from the N terminal of SMPD2. Peptide sequence RQKLSPTYPAAHHFRSGIIGSGLCVFSKHPIQELTQHIYTLNGYPYMIHH The peptide sequence for this immunogen was taken from within the described region.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6610
Test Specificity Expected identity based on immunogen sequence: Human: 100%; Canine: 85%; Equine: 85%; Bovine: 78%; Pig: 78%; Mouse: 76%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.