Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUDT16L1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$480.74
Specifications
| Antigen | NUDT16L1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP155432
![]() |
Novus Biologicals
NBP155432 |
100 μL |
Each for $480.74
|
|
|||||
NBP15543220
![]() |
Novus Biologicals
NBP15543220UL |
20 μL | N/A | N/A | N/A | ||||
Description
NUDT16L1 Polyclonal specifically detects NUDT16L1 in Human samples. It is validated for Western Blot.Specifications
| NUDT16L1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| MGC11275, nudix (nucleoside diphosphate linked moiety X)-type motif 16-like 1, protein syndesmos, SDOSNUDT16-like protein 1 | |
| NUDT16L1 | |
| Primary | |
| 23 kDa |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Q9BRJ7 | |
| 84309 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title