Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Obsidian Specimen
SCI_ED

Catalog No. S26587 Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm

Obsidian Specimen, also known as volcanic glass, forms from lava that cools almost immediately upon eruption.

  • Volcanic glass formed through nearly instantaneous quenching of lava
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Obsidian shares many physical properties with its major chemical component, quartz, despite looking dissimilar
  • Despite being dark in color, obsidian is made up of microcrystalline quartz, demonstrated by sharing hardness and conchoidal fracture often exhibited by quartz
  • For use in classroom identification labs and exercises

Specifications

Product Type Igneous Rock, Obsidian
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.