Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCT4 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1002379SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002379SS 0.025 mL
NB1002379 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1002379SS Supplier Novus Biologicals Supplier No. NB1002379SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 18 publications

44473 Polyclonal specifically detects 44473 in Human, Mouse, Rat, Bovine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen OCT4
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot reported by customer review, Flow Cytometry 1:50-1:200, Immunohistochemistry 1:200. Use reported in scientific literature (PMID 30726734), Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q01860
Gene Alias class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein
Gene Symbols POU5F1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human OCT4 protein (between residues 100-200) [UniProt# Q01860]
Molecular Weight of Antigen 40 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, Embryonic Stem Cell Markers, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5460
Test Specificity Human, monkey, rat, mouse. Immunogen has 93% homology to cow, feline and pig. Customer feedback suggests that this antibody does not give good results in IHC-P on feline tissues.
Target Species Human, Mouse, Rat, Primate
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.