Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCT4 Antibody (CL7621), Novus Biologicals™
SDP

Catalog No. NB239258 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB239258 100 μg
NB239265 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB239258 Supplier Novus Biologicals Supplier No. NBP321158100UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OCT4 Monoclonal antibody specifically detects OCT4 in Human samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen OCT4
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone CL7621
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 2-10 ug/ml
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein
Host Species Mouse
Immunogen This antibody was developed against a Synthetic Peptide of Q01860. Actual immunogen sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5460
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.