Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Octyl Sepharose™ 4 Fast Flow Chromatography Media

Catalog No. p-4592264 Shop All Cytiva Products
Click to view available options
Quantity:
200 mL

Aliphatic hydrophobic interaction chromatography (HIC) medium for capture and intermediate purification of larger proteins

  • HIC ligands are coupled to monosacharide units via their corresponding glycidylethers, giving matrices without charges and stable etherbonds between ligands and agarose
  • Octyl derivative adds different and complementary selectivity to Sepharose Fast Flow HIC product family
  • Resin based on cross-linked 4% agarose
  • Supported for industrial applications and well established in approved processes
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 mL
For Use With (Application) For Capture and Intermediate PurifiCatIon of Larger Proteins
Stability Stable in commonly used aqueous Buffers: 1 M NaOH, 30% Isopropanol, 70% Ethanol, 6 M Guanidine-Hydrochloride, 30% Acetonitrile, 1 mM HCl
Storage Requirements 4°C to 30°C
Product Type Octyl Sepharose 4 Fast Flow Media
Flow Rate 150 to 250 cm/Hoμr
Particle Size 45 to 165 μm
Height (English) Bed 9.84 in.
Height (Metric) Bed 25 cm
Max. Pressure 100 kPa
Matrix 4% Cross-Linked Agarose
pH Range 2 to 14
Storage Buffer 20% Ethanol
Ligand Type Octyl
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.