Learn More
Octylmagnesium bromide, 2M solution in diethyl ether,AcroSeal™
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 35.0°C |
| Color | Yellow to Brown |
| CAS Min % | 42.99, 50.22 |
| CAS Max % | 49.78, 57.01 |
| Physical Form | Liquid |
| Quantity | 800 mL |
| Linear Formula | CH3(CH2)7MgBr |
| Flash Point | −35°C |
| Formula Weight | 217.43 |
| Concentration or Composition (by Analyte or Components) | 1.90 to 2.20M |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause drowsiness or dizziness. Causes severe skin burns and eye damage. Harmful if swallowed. Extremely flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsi |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.