Learn More
Eisco™ Oligoclase Mineral Specimen
SCI_ED
Catalog No. p-7203000 Shop All Eisco Products
Click to view available options
Quantity:
1/Ea.
12/Pk.
Oligoclase is a feldspar with high levels of sodium.
  • Correlates well to Next Generation Science Standards Evidence
  • Standard hardness of 6
  • Exhibits two planes of cleavage
  • Perfect size for use in classroom identification labs and exercises
Specifications
Product Type Mineral
Grade K to 16
Quantity 1/Ea.
Specimen Size 2 to 3 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.