Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Olvanil
GREENER_CHOICE

Catalog No. p-4785284
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent vanilloid receptor agonist

Potent vanilloid receptor agonist (pEC50 values are 8.1 and 7.7 at rat and human VR1 receptors respectively). Also blocks anandamide uptake (IC50 = 9 μM) and may bind to CB1 cannabinoid receptors. Antinociceptive following systemic administration.

Chemical Identifiers

CAS 58493-49-5
Molecular Formula C26H43NO3

Specifications

Quantity 10 mg
Formula Weight 417.63
Purity >98%
Molecular Weight (g/mol) 417.634
Chemical Name or Material Olvanil
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.