Learn More
Description
Specifications
Specifications
| Antigen | OR10A4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | HP2, hP2 olfactory receptor, JCG5, olfactory receptor 10A4, olfactory receptor OR11-87, olfactory receptor, family 10, subfamily A, member 4, olfactory receptor, family 10, subfamily A, member 4 pseudogene, Olfactory receptor-like protein JCG5, OR10A4P |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR10A4 (NP_997069). Peptide sequence TAILTYFRPQSSASSESKKLLSLSSTVVTPMLNPIIYSSRNKEVKAALKR |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
