Learn More
Description
Specifications
Specifications
| Antigen | p21/CIP1/CDKN1A |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Monoclonal |
| Clone | HJ21 |
| Conjugate | Alexa Fluor 532 |
| Dilution | Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | CAP20cyclin-dependent kinase inhibitor 1, CDK-interacting protein 1, CDKN1melanoma differentiation associated protein 6, CIP1WAF1CDK-interaction protein 1, cyclin-dependent kinase inhibitor 1A (p21, Cip1), MDA6, MDA-6, Melanoma differentiation-associated protein 6, p21, p21CIP1, p21Cip1/Waf1, PIC1, SDI1DNA synthesis inhibitor, wild-type p53-activated fragment 1 |
| Gene Symbols | CDKN1A |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
