Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p23/PTGES3 Antibody (OTI2D2) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP273186 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273186 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273186 Supplier Novus Biologicals Supplier No. NBP273186
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p23/PTGES3 Monoclonal specifically detects p23/PTGES3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen p23/PTGES3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2D2
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunohistochemistry 1:150
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias cPGESp23, Cytosolic prostaglandin E2 synthase, Hsp90 co-chaperone, P23cytosolic prostaglandin E synthase, Progesterone receptor complex p23, prostaglandin E synthase 3, prostaglandin E synthase 3 (cytosolic), TEBPEC 5.3.99.3, Telomerase-binding protein p23, unactive progesterone receptor, 23 kD
Gene Symbols PTGES3
Host Species Mouse
Immunogen Full length human recombinant protein of human PTGES3 (NP_006592) produced in E.coli.
Molecular Weight of Antigen 18.5 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 10728
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.