Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2X7/P2RX7 Antibody (Hano43) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018511 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018511 0.025 mg
NBP14089401 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018511 Supplier Novus Biologicals Supplier No. NBP1408940.025MG
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody has been used in 2 publications

P2X7/P2RX7 Monoclonal specifically detects P2X7/P2RX7 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen P2X7/P2RX7
Applications Immunocytochemistry, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone Hano43
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:25-1:200, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Formulation PBS (pH 7.4) with 0.09% Sodium Azide
Gene Accession No. Q9Z1M0
Gene Alias ATP receptor, MGC20089, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A
Gene Symbols P2RX7
Host Species Rat
Immunogen A P2X7-expression construct and a final boost with P2X7-transfected HEK cells.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5027
Test Specificity This antibody recognises the P2X purinoceptor 7, also known as P2X7. The P2X7 ATP receptor has a distinctive long C-terminal tail with multiple potential protein and lipid interaction motifs and is highly polymorphic. It is a cation selective ion channel that opens up on binding of extracellular ATP. Sustained activation by extracellular ATP results in the formation of a reversible pore in the plasma membrane that is permeable to hydrophilic solutes of up to 900 Da. Once a pore is opened massive upset of cytoplasmic ion homeostasis occurs and the pore stays open as long as it is bound by ATP. Should ATP stimulation continue the cell will become irreversibly damaged and die. P2X7 plays a key role in the maturation and release of IL-1 and other IL-1 family members during inflammation. As such, P2X7 blockers might be useful as anti-inflammatory agents.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.