Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PABPC1L2A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | PABPC1L2A |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PABPC1L2A Polyclonal specifically detects PABPC1L2A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PABPC1L2A | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
340529 | |
This antibody was developed against a Recombinant Protein corresponding to amino acids:KSHKEREAERGAWARQSTSADVKDFEEDTDEEATLR | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
MGC168104, PABPC1L2, PABPC1L2B, poly(A) binding protein, cytoplasmic 1-like 2A, polyadenylate-binding protein 1-like 2, RBM32A, RBM32B, RNA binding motif protein 32A, RNA-binding motif protein 32, RNA-binding protein 32 | |
PABPC1L2A | |
IgG | |
Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title