Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAK1, p Ser204 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $816.50
Specifications
| Antigen | PAK1 (p Ser204) |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
PAK1 Polyclonal specifically detects PAK1 in Human samples. It is validated for Western Blot.Specifications
| PAK1 (p Ser204) | |
| Western Blot 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Breast Cancer, Cancer, Neuroscience, Phospho Specific, Signal Transduction | |
| PBS and 50% Glycerol with 0.05% Sodium Azide | |
| alpha-PAK, EC 2.7.11, EC 2.7.11.1, MGC130000, MGC130001, p21 protein (Cdc42/Rac)-activated kinase 1, p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog, yeast), p21/Cdc42/Rac1-activated kinase 1 (yeast Ste20-related), p21-activated kinase 1, p65-PAK, PAK-1, PAKalpha, serine/threonine-protein kinase PAK 1, STE20 homolog, yeast | |
| PAK1 | |
| IgG | |
| Affinity Purified | |
| Recognizes PAK1 phosphorylated on Ser199 and Ser204 (human)/Ser198 and Ser203 (mouse). Immunogen sequence is higly homolgous (greater than 90%) with PAK2 and PAK3. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Q13153 | |
| 5058 | |
| Amino Acids encompassing and including phosphorylated Ser199 and Ser204 of human PAK1. Sequence is highly homologous (greater than 90%) to PAK2 and PAK3. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 65 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title