Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAPD4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | PAPD4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP16906020
![]() |
Novus Biologicals
NBP16906020UL |
20 μL |
Each for $206.00
|
|
|||||
NBP169060
![]() |
Novus Biologicals
NBP169060 |
100 μL |
Each for $487.50
|
|
|||||
Description
PAPD4 Polyclonal specifically detects PAPD4 in Rat samples. It is validated for Western Blot.Specifications
PAPD4 | |
Polyclonal | |
Rabbit | |
Q5U315 | |
167153 | |
IgG | |
56 kDa |
Western Blot | |
Unconjugated | |
RUO | |
EC 2.7.7.19, FLJ38499, GLD2TUTase 2, hGLD-2, PAP associated domain containing 4, PAP-associated domain-containing protein 4, poly(A) RNA polymerase GLD2, Terminal uridylyltransferase 2 | |
Synthetic peptides corresponding to Papd4 (PAP associated domain containing 4) The peptide sequence was selected from the C terminal of Papd4. Peptide sequence YICVEEPFDGTNTARAVHEKQKFDMIKDQFLKSWQRLKNKRDLNSVLPLR. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title