Learn More
Applied Biosystems™ Path-ID™ qPCR Master Mix
Description
Ambion™ Path-ID™ qPCR Master Mix is optimized for targets of importance in animal health and to function in the presence of PCR inhibitors. The mix is supplied in one tube suitable for 100 reactions. With the exception of PCR primers and probes for your target, the master mix contains everything you need for real-time PCR, including:
- AmpliTaq Gold™ ultrapure thermostable DNA polymerase designed for automatic hot start, to minimize nonspecific PCR products
- Optimized buffer and dNTP concentration, for reliable PCR
- Passive internal reference, based on proprietary ROX™ dye, for precise data analysis.
Specifications
Specifications
| Detection Method | Primer-probe |
| Form | Liquid |
| Format | Tube |
| PCR Method | qPCR |
| Polymerase | AmpliTaq Gold DNA Polymerase |
| Reaction Speed | Fast or Standard |
| For Use With (Equipment) | 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode |
| Passive Reference Dye | ROX (Pre-mixed) |
| Product Line | Ambion, Path-ID |
| Purity or Quality Grade | UP (Ultra Pure) |
| Show More |
For Veterinary Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.