Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCDH15 Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $511.50
Specifications
Antigen | PCDH15 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119378
|
Invitrogen™
PA5119378 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121704
|
Invitrogen™
PA5121704 |
20 μL |
Each for $237.00
|
|
|||||
Description
PCDH15 Polyclonal Antibody for Western Blot, IHC (P)
PCDH15 is a member of the cadherin superfamily. Family members encode integral membrane proteins that mediate calcium-dependent cell-cell adhesion. PCDH15 consists of a signal peptide, 11 extracellular calcium-binding domains, a transmembrane domain and a unique cytoplasmic domain. It plays an essential role in maintenance of normal retinal and cochlear function. Mutations in this gene have been associated with hearing loss, which is consistent with its location at the Usher syndrome type 1F (USH1F) critical region on chromosome 10.Specifications
PCDH15 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Mouse | |
PCDH15 | |
Ames waltzer; av; BB078305; cadherin-related family member 15; CDHR15; DFNB23; DKFZp667A1711; ENSMUSG00000046980; Gm9815; nmf19; Pcdh15; protocadherin 15; protocadherin 15 CD2; protocadherin 15 CD3 isoform; protocadherin related 15; protocadherin-15; protocadherin-15-CD2; protocadherin-related 15; RP11-449J3.2; USH1F | |
PCDH15 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q99PJ1 | |
11994 | |
Recombinant protein Protocadherin 15. The antigen corresponds to amino acid range 45-270 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title